Cutting board
Lieferant: KLINIPATH
060.012.119
KPXX060.012.119EA 353 EUR
KPXX060.012.119
Cutting board
Präparierbretter
The cutting board is completely made of PE.
The top thickness is approx. 20 mm. The board includes 4 small feet of 25 mm height and 15 mm diameter.
Bitte beachten Not suitable for thermal sterilisation.