Bromphenolblau 0,1% in Ethanol

Lieferant: Bernd Kraft
Gefahr

Synonyme: 3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

03018.3000 03018.1000 03018.0000 03018.2000
KRAF03018.3000EA 130 EUR
KRAF03018.3000 KRAF03018.1000 KRAF03018.0000 KRAF03018.2000
Bromphenolblau 0,1% in Ethanol
Bromphenolblau
Formel: C₁₉H₁₀Br₄O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00005875
CAS-Nummer: 115-39-9
EINECS: 204-086-2
UN: 1170
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR