Neßler-Reagenz

Lieferant: Bernd Kraft
Gefahr

Synonyme: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

03036.1000
KRAF03036.1000EA 64 EUR
KRAF03036.1000
Neßler-Reagenz
Neßler-Reagenz
Molecular Weight: 786,4 g/mol
Siedepunkt: 356,6 °C (1013 hPa)
Schmelzpunkt: –38,9 °C
Dichte: 13,6 g/cm³ (20 °C)
MDL: MFCD00049656
CAS-Nummer: 7783-33-7
EINECS: 231-990-4
UN: 2922
ADR: 8,II
Merck Index: 12,07862

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR