Natriumthiosulfat 0.1 mol/l (0.1 N) aqueous solution
Lieferant: Bernd Kraft
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
03067.3000
03067.4000
03067.5000
03067.5300
03067.6000
03067.6400
03067.7000
03067.5400
KRAF03067.3000EA
24.4
EUR
KRAF03067.3000
KRAF03067.4000
KRAF03067.5000
KRAF03067.5300
KRAF03067.6000
KRAF03067.6400
KRAF03067.7000
KRAF03067.5400
Natriumthiosulfat 0.1 mol/l (0.1 N) aqueous solution
Natriumthiosulfat
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 Schmelzpunkt: 48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...