Buffer solutions, ready to use
Lieferant: Bernd Kraft
03083.6400
03084.6400
03084.7900
03086.6400
03087.6400
03088.6400
03089.6400
03093.6400
03093.3000
03081.3000
03083.3000
03086.3000
03088.3000
03089.3000
03200.5000
03081.2000
03081.5000
03081.6400
03082.2000
03082.3000
03082.5000
03082.6400
03083.2000
03083.5000
03083.6000
03085.2000
03085.3000
03086.2000
03086.5000
03086.6000
03087.2000
03087.3000
03087.5000
03088.2000
03088.5000
03089.2000
03089.5000
03090.2000
03090.3000
03090.5000
03090.6400
03093.2000
03096.3000
03080.3000
03080.2000
03080.3060
03080.5000
03081.3060
03081.6000
03082.3060
03083.3060
03084.2000
03084.3000
03084.3060
03084.5000
03085.3060
03085.6000
03086.3060
03087.3060
03088.3060
03089.3060
03090.3060
03093.3060
03096.3060
03397.3000
03397.3060
03080.6400
03080.5400
03080.6000
03081.5400
03082.5400
03082.6000
03083.5400
03084.5400
03084.6000
03085.5000
03085.5400
03085.6400
03086.5400
03087.5400
03087.6000
03088.5400
03088.6000
03089.5400
03089.6000
03090.5400
03090.6000
03093.5000
03093.5400
03093.6000
03096.2000
03096.5000
03096.5400
03096.6000
03096.6400
03397.2000
03397.5000
03397.5400
03397.6000
03397.6400
03200.2000
03200.3060
03200.5400
03200.6000
03200.6400
KRAF03083.6400EA
181
EUR
KRAF03083.6400
KRAF03084.6400
KRAF03084.7900
KRAF03086.6400
KRAF03087.6400
KRAF03088.6400
KRAF03089.6400
KRAF03093.6400
KRAF03093.3000
KRAF03081.3000
KRAF03083.3000
KRAF03086.3000
KRAF03088.3000
KRAF03089.3000
KRAF03200.5000
KRAF03081.2000
KRAF03081.5000
KRAF03081.6400
KRAF03082.2000
KRAF03082.3000
KRAF03082.5000
KRAF03082.6400
KRAF03083.2000
KRAF03083.5000
KRAF03083.6000
KRAF03085.2000
KRAF03085.3000
KRAF03086.2000
KRAF03086.5000
KRAF03086.6000
KRAF03087.2000
KRAF03087.3000
KRAF03087.5000
KRAF03088.2000
KRAF03088.5000
KRAF03089.2000
KRAF03089.5000
KRAF03090.2000
KRAF03090.3000
KRAF03090.5000
KRAF03090.6400
KRAF03093.2000
KRAF03096.3000
KRAF03080.3000
KRAF03080.2000
KRAF03080.3060
KRAF03080.5000
KRAF03081.3060
KRAF03081.6000
KRAF03082.3060
KRAF03083.3060
KRAF03084.2000
KRAF03084.3000
KRAF03084.3060
KRAF03084.5000
KRAF03085.3060
KRAF03085.6000
KRAF03086.3060
KRAF03087.3060
KRAF03088.3060
KRAF03089.3060
KRAF03090.3060
KRAF03093.3060
KRAF03096.3060
KRAF03397.3000
KRAF03397.3060
KRAF03080.6400
KRAF03080.5400
KRAF03080.6000
KRAF03081.5400
KRAF03082.5400
KRAF03082.6000
KRAF03083.5400
KRAF03084.5400
KRAF03084.6000
KRAF03085.5000
KRAF03085.5400
KRAF03085.6400
KRAF03086.5400
KRAF03087.5400
KRAF03087.6000
KRAF03088.5400
KRAF03088.6000
KRAF03089.5400
KRAF03089.6000
KRAF03090.5400
KRAF03090.6000
KRAF03093.5000
KRAF03093.5400
KRAF03093.6000
KRAF03096.2000
KRAF03096.5000
KRAF03096.5400
KRAF03096.6000
KRAF03096.6400
KRAF03397.2000
KRAF03397.5000
KRAF03397.5400
KRAF03397.6000
KRAF03397.6400
KRAF03200.2000
KRAF03200.3060
KRAF03200.5400
KRAF03200.6000
KRAF03200.6400
Buffer solutions, ready to use
Puffer
pH-Puffer
- NIST traceable
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...