Natriumthiosulfat 0.02 mol/l (0.02 N) aqueous solution
Lieferant: Bernd Kraft
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
03166.5000
03166.3000
03166.5400
03166.6000
03166.6400
KRAF03166.5000EA
140
EUR
KRAF03166.5000
KRAF03166.3000
KRAF03166.5400
KRAF03166.6000
KRAF03166.6400
Natriumthiosulfat 0.02 mol/l (0.02 N) aqueous solution
Natriumthiosulfat
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 Schmelzpunkt: 48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...