Phenolphthalein 0,2% in Methanol

Lieferant: Bernd Kraft
Gefahr

03783.3700 03783.1700 03783.2700 03783.4700
KRAF03783.3700EA 55.4 EUR
KRAF03783.3700 KRAF03783.1700 KRAF03783.2700 KRAF03783.4700
Phenolphthalein 0,2% in Methanol
Phenolphthalein
Formel: C₂₀H₁₄O₄
Dichte: 0,8 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005913
CAS-Nummer: 77-09-8
EINECS: 201-004-7
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR