Natriumthiosulfat 0.0394 mol/l (0.0394 N) aqueous solution
Lieferant: Bernd Kraft
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
05443.3000
05443.5000
05443.5400
05443.6000
KRAF05443.3000EA
250
EUR
KRAF05443.3000
KRAF05443.5000
KRAF05443.5400
KRAF05443.6000
Natriumthiosulfat 0.0394 mol/l (0.0394 N) aqueous solution
Natriumthiosulfat
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 Schmelzpunkt: 48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...