Bathocuproin 0,5 g/l in wässriger Lösung
Lieferant: Bernd Kraft
Synonyme:
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
05846.6000
KRAF05846.6000EA
0
EUR
KRAF05846.6000
Bathocuproin 0,5 g/l in wässriger Lösung
Bathocuproin
Formel:
C₂₆H₂₀N₂ Molecular Weight: 360,46 g/mol |
MDL:
MFCD00004972 CAS-Nummer: 4733-39-5 EINECS: 225-240-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...