Glycerin 86 - 88% ≥86-88%

Lieferant: Bernd Kraft

Synonyme: 1,2,3-Propantriol, Glycerol

07092.3000 07092.5000
KRAF07092.3000EA 41.4 EUR
KRAF07092.3000 KRAF07092.5000
Glycerin 86 - 88% ≥86-88%
Glycerin
Formel: HOCH₂CH(OH)CH₂OH
Lagertemperatur: Raumtemperatur
MDL: MFCD00004722
CAS-Nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR