Hydroxylammoniumchlorid 4,5% in wässriger Lösung
Lieferant: Bernd Kraft
Synonyme:
Hydroxylamin-Hydrochlorid, Chlorhydrathydroxylamin, Hydroxylamin Hydrochlorid, Hydroxylamin HCl
11512.4000
KRAF11512.4000EA
0
EUR
KRAF11512.4000
Hydroxylammoniumchlorid 4,5% in wässriger Lösung
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Molecular Weight: 69,49 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN: 1789 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...