Pararosaniliniumchlorid Reagent R Reag. Ph. Eur.

Lieferant: Bernd Kraft
Gefahr

Synonyme: Basic Red 9, Fuchsin basisch Hydrochlorid, 4,4'-(4-Iminocyclohexa-2,5-dienylidenmethylen)dianilinhydrochlorid, 4,4',4''-Triaminotrityliumchlorid, 4,4′-(4-Iminocyclohexa-2,5-dienylidenmethylen)dianilinhydrochlorid, Pararosanilin, Parafuchsin

13355.1700
KRAF13355.1700EA 232 EUR
KRAF13355.1700
Pararosaniliniumchlorid Reagent R Reag. Ph. Eur.
Pararosaniliniumchlorid
Formel: C₁₉H₁₈ClN₃
Molecular Weight: 323,82 g/mol
MDL: MFCD00001657
CAS-Nummer: 569-61-9
UN: 1789
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR