Ammoniumthiocyanat, reinst
Lieferant: Bernd Kraft
|
Achtung
|
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
15200.3600
15200.5600
15200.2600
15200.4600
15200.6600
KRAF15200.3600EA
69.5
EUR
KRAF15200.3600
KRAF15200.5600
KRAF15200.2600
KRAF15200.4600
KRAF15200.6600
Ammoniumthiocyanat, reinst
Ammoniumthiocyanat
Formel:
NH₄SCN Schmelzpunkt: 150 °C Dichte: 1,305 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...