Calciumsulfat Dihydrat, extra_pure
Lieferant: Bernd Kraft
Synonyme:
Pflaster Paris
15219.3600
15219.5600
15219.1600
15219.6600
KRAF15219.3600EA
42
EUR
KRAF15219.3600
KRAF15219.5600
KRAF15219.1600
KRAF15219.6600
Calciumsulfat Dihydrat, extra_pure
Calciumsulfat Dihydrat
Formel:
CaSO₄·2H₂O Molecular Weight: 172.17 g/mol Dichte: 2.32 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149625 CAS-Nummer: 10101-41-4 EINECS: 231-900-3 Merck Index: 13,01711 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...