Dimethylglyoxim 1% in Ethanol 96% (v/v)

Lieferant: Bernd Kraft

16495.3700
KRAF16495.3700EA 114 EUR
KRAF16495.3700
Dimethylglyoxim 1% in Ethanol 96% (v/v)
Dimethylglyoxim
Formel: CH₃C(=NOH)C(=NOH)CH₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00002117
CAS-Nummer: 95-45-4
EINECS: 202-420-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR