Bromthymolblau

Lieferant: Bernd Kraft

Synonyme: 3′,3′′-Dibromthymolsulfonphthalein, BTB

17778.0801
KRAF17778.0801EA 44.6 EUR
KRAF17778.0801
Bromthymolblau
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
Molecular Weight: 624.39 g/mol
Schmelzpunkt: 204…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR