Kaliumchlorid 50 g/l aqueous solution zA
Lieferant: Bernd Kraft
Synonyme:
Potassiumchloride
18435.2000
18435.3000
18435.5020
KRAF18435.2000EA
0
EUR
KRAF18435.2000
KRAF18435.3000
KRAF18435.5020
Kaliumchlorid 50 g/l aqueous solution zA
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 Schmelzpunkt: 773 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...