Bromphenolblau Natriumsalz
Lieferant: Bernd Kraft
20091.0801
20091.0802
20091.0803
KRAF20091.0801EA
35.8
EUR
KRAF20091.0801
KRAF20091.0802
KRAF20091.0803
Bromphenolblau Natriumsalz
Bromphenolblau Natriumsalz
Formel:
C₁₉H₉Br₄NaO₅S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013793 CAS-Nummer: 34725-61-6 EINECS: 252-170-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...