Buffer indicator tablets for water hardness
Lieferant: Bernd Kraft
20506.3601
KRAF20506.3601EA
109
EUR
KRAF20506.3601
Buffer indicator tablets for water hardness
Umwelt-Prüfkits
Indicator buffer tablets used for the determination of the water hardness with EDTA.