5-(4-(Dimethylamino)benzyliden)rhodanin analytisches Reagens

Lieferant: Bernd Kraft

20654.0900
KRAF20654.0900EA 78.3 EUR
KRAF20654.0900
5-(4-(Dimethylamino)benzyliden)rhodanin analytisches Reagens
5-(4-(Dimethylamino)benzyliden)rhodanin
Formel: C₁₂H₁₂N₂OS₂
MDL: MFCD00064857
CAS-Nummer: 536-17-4
EINECS: 208-625-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR