Bromthymolblau 0,5 g/l in wässriger Lösung
Lieferant: Bernd Kraft
Synonyme:
3′,3′′-Dibromthymolsulfonphthalein, BTB
20682.0700
KRAF20682.0700EA
111
EUR
KRAF20682.0700
Bromthymolblau 0,5 g/l in wässriger Lösung
Bromthymolblau
Formel:
C₂₇H₂₈Br₂O₅S Molecular Weight: 624.39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005872 CAS-Nummer: 76-59-5 EINECS: 200-971-2 Merck Index: 13,01430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...