Kernechtrot

Lieferant: Bernd Kraft

Synonyme: Natrium-1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracen-3-sulfonat, Heliechtrubin

21481.0802
KRAF21481.0802EA 0 EUR
KRAF21481.0802
Kernechtrot
Kernechtrot
Formel: C₁₄H₈NNaO₇S
Molecular Weight: 357,28 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078493
CAS-Nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR