Chlorwasserstoff Ph. Eur.

Lieferant: Bernd Kraft

Synonyme: Chlorwasserstoff

26780.3000
KRAF26780.3000EA 134 EUR
KRAF26780.3000
Chlorwasserstoff Ph. Eur.
Chlorwasserstoff
Formel: HCl
Molecular Weight: 36.46 g/mol
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
UN: 1050
ADR: 2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR