Nitrate/ammonium standard solution
Lieferant: Bernd Kraft
27278.2000
KRAF27278.2000EA
102
EUR
KRAF27278.2000
Nitrate/ammonium standard solution
Standards
Anionische Standards
Nitrat-Standards
Solution with 100 mg each NH₄-N + NO₃-N/l KNO₃ + (NH₄)₂SO₄ in water for TNb determination.