Kristallviolett-Lösung R Reag. Ph. Eur. chapter 4.1.1

Lieferant: Bernd Kraft
Gefahr

Synonyme: Hexamethyl-p-rosaniliniumchlorid, Hexamethylen-p-rosanilinchlorid, blaues Pyoktanin, Basic Violet 3, Hexamethylviolett, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid, Methylviolett 10B, Methylrosanilin, Methylrosanilium, 4,4',4''-Tri(dimethylamino)trityliumchlorid, CI 42555, Methylrosaniliumchlorid, Gentianaviolett

27773.0700
KRAF27773.0700EA 121 EUR
KRAF27773.0700
Kristallviolett-Lösung R Reag. Ph. Eur. chapter 4.1.1
Kristallviolett
Formel: C₂₅H₃₀ClN₃
Molecular Weight: 407,99 g/mol
MDL: MFCD00011750
CAS-Nummer: 548-62-9
UN: 2789
ADR: 3.8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR