Hazardous substance labels
Lieferant: KROSCHKE
27478
KROS27478EA 208 EUR
KROS27478
Hazardous substance labels
Etiketten
Hazard symbol label with question mark, used for unverified research material.
  • Self-adhesive
  • Resistant to water, alkalis and oil
  • Temperature resistant from –40 to +80 °C