Spectroscope, Kirchhoff-Bunsen
Lieferant: A.KRÜSS Optronic
1701
KRUE1701EA 1760 EUR
KRUE1701
Spectroscope, Kirchhoff-Bunsen
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
Spectroscope suitable for qualitative analysis as well as measurement of emission and absorption spectra.
  • Moveable observation tube with lock-screw ocular
  • Fixed slit tube with variable slit objective: 18 / 160 mm
  • Fixed scale tube with scale of 200 divisions ocular: 18 / 90 mm
  • Angle dispersion C-F = 2°