Fully automatic polarimeter
Lieferant: A.KRÜSS Optronic
P3000
KRUEP3000EA 0 EUR
KRUEP3000
Fully automatic polarimeter
Polarimeter
This digital polarimeter is designed to measure optically active liquids. The measuring time is one second, regardless of the rotation angle of the sample.
  • Two decimal point accuracy
  • Time savings with about one second measurement duration
  • Measured values displayed as optical rotaion or in ICUMSA sugar scale