Gentamicinsulfat solution_SC 10 mg/ml

Lieferant: Biowest

Synonyme: Gentamicin Sulfatsalz

L0011-010 L0011-100
L0011-010EA 14 EUR
L0011-010 L0011-100
Gentamicinsulfat solution_SC 10 mg/ml
Gentamicinsulfat

Gentamicin sulfate acts against gram-negative, gram-positive bacteria and mycoplasma.


  • Storage temperature of –20 °C
  • pH of 4,5±0,3
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml (<1 ng/ml)


Used in cell culture applications at 5 ml/l. This concentration is for tissue culture media containing serum, serum-free media generally require lower concentration.


Bitte beachten This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

MDL: MFCD00270181
CAS-Nummer: 1405-41-0
Merck Index: 13,04403

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR