1-(Trifluoracetyl)imidazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluoro-1-(1H-imidazol-1-yl)ethanone
L00135.09
L00135.18
L00135.09EA
83.3
EUR
L00135.09
L00135.18
1-(Trifluoracetyl)imidazol ≥98%
1-(Trifluoracetyl)imidazol
Purity: 98+%
CAS No.: 1546-79-8
Molecular Formula: C5H3F3N2O
Formula Weight: 164.09
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4240
Odour: Pungent
Boiling Point: 45-46°/14mm
Beilstein Reference: 608904
UN No.: 1993
MDL No.: MFCD00014501
Formel:
C₅H₃F₃N₂O Molecular Weight: 164.09 g/mol Siedepunkt: 45…46 °C (14 mmHg) Dichte: 1.441 Flammpunkt: 45 °C (113 °F) |
MDL:
MFCD00014501 CAS-Nummer: 1546-79-8 EINECS: 216-282-5 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...