1-(Trifluoracetyl)imidazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2,2-Trifluoro-1-(1H-imidazol-1-yl)ethanone

L00135.09 L00135.18
L00135.09EA 83.3 EUR
L00135.09 L00135.18
1-(Trifluoracetyl)imidazol ≥98%
1-(Trifluoracetyl)imidazol

Purity: 98+% CAS No.: 1546-79-8 Molecular Formula: C5H3F3N2O Formula Weight: 164.09 Storage Temperature: 2°C to 8°C Flash Point: 45°(113°F) Refractive Index: 1.4240 Odour: Pungent Boiling Point: 45-46°/14mm Beilstein Reference: 608904 UN No.: 1993 MDL No.: MFCD00014501

Formel: C₅H₃F₃N₂O
Molecular Weight: 164.09 g/mol
Siedepunkt: 45…46 °C (14 mmHg)
Dichte: 1.441
Flammpunkt: 45 °C (113 °F)
MDL: MFCD00014501
CAS-Nummer: 1546-79-8
EINECS: 216-282-5
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR