G418 -Lösung

Lieferant: Biowest

Synonyme: G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol

L0015-020 L0015-100
L0015-020EA 81 EUR
L0015-020 L0015-100
G418 -Lösung
G418

Antibiotic G418 is an aminoglycoside antibiotic similar in structure to gentamycin. It exhibits toxicity towards both eukaryotic and prokaryotic cells. The optimal concentration for selection and maintenance must be determined for each cell line.


  • Clear to very slightly hazy liquid
  • Shelf life of 24 months
  • Storage temperature of –20 °C


Bitte beachten This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Formel: C₂₀H₄₀N₄O₁₀
Molecular Weight: 496.552 g/mol
MDL: MFCD30541267
CAS-Nummer: 49863-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR