(Ethoxycarbonylmethyl)dimethylsulphonium bromide 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-Ethoxy-2-oxoethyl)(dimethyl)sulphonium bromide
L00266.09
L00266.18
L00266.09EA
67
EUR
L00266.09
L00266.18
(Ethoxycarbonylmethyl)dimethylsulphonium bromide 98%
(Ethoxycarbonylmethyl)dimethylsulphonium bromide
Purity: 98%
CAS No.: 5187-82-6
Molecular Formula: CH3CH2O2CCH2S(CH3)2Br
Formula Weight: 229.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 80° dec.
Beilstein Reference: 4161472
MDL No.: MFCD00031604
Formel:
CH₃CH₂O₂CCH₂S(CH₃)₂Br Molecular Weight: 229.14 g/mol Schmelzpunkt: ∼ 80 °C (Zers.) |
MDL:
MFCD00830557 CAS-Nummer: 5187-82-6 EINECS: 225-968-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...