N-Methylmaleimid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L00437.06 L00437.14
L00437.06EA 53 EUR
L00437.06 L00437.14
N-Methylmaleimid ≥98%
N-Methylmaleimid

Purity: 98+% CAS No.: 930-88-1 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Storage Temperature: 2°C to 8°C Odour: Odourless Beilstein Reference: 108550 UN No.: 2928 MDL No.: MFCD00005508

Formel: C₅H₅NO₂
Siedepunkt: 194 °C (1013 hPa)
Schmelzpunkt: 94…96 °C
MDL: MFCD00005508
CAS-Nummer: 930-88-1
EINECS: 213-226-1
UN: 2928
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR