Methyl (methylsulphinyl)methyl sulphide 97%

Lieferant: Thermo Fisher Scientific

Synonyme: FAMSO, Methyl((methylsulphinyl)methyl)sulphane, Methyl (Methylthio)methyl sulphoxide, Formaldehyde Dimethyl Dithioacetal S-Oxide

L00510.09
L00510.09EA 41.3 EUR
L00510.09
Methyl (methylsulphinyl)methyl sulphide 97%
Methyl (methylsulphinyl)methyl sulphide

Purity: 97% CAS No.: 33577-16-1 Molecular Formula: C3H8OS2 Formula Weight: 124.22 Flash Point: 110°(230°F) Refractive Index: 1.5520 Boiling Point: 222-226° Beilstein Reference: 906789 MDL No.: MFCD00002091

Formel: C₃H₈OS₂
Molecular Weight: 124,22 g/mol
Siedepunkt: 222…226 °C
Dichte: 1,222
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002091
CAS-Nummer: 33577-16-1
EINECS: 251-577-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR