4-Methoxybenzyl carbazate 97%

Lieferant: Thermo Fisher Scientific
Achtung

L00541.06
L00541.06EA 49.5 EUR
L00541.06
4-Methoxybenzyl carbazate 97%
4-Methoxybenzyl carbazate

Purity: 97% CAS No.: 18912-37-3 Molecular Formula: C9H12N2O3 Formula Weight: 196.21 Melting Point: 75-77° Beilstein Reference: 2052164 MDL No.: MFCD00042817

Formel: C₉H₁₂N₂O₃
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042817
CAS-Nummer: 18912-37-3
EINECS: 242-664-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR