1H-Benzimidazol-2-ylacetonitril 99%

Lieferant: Thermo Fisher Scientific
Achtung

L00827.09 L00827.18
L00827.09EA 82.2 EUR
L00827.09 L00827.18
1H-Benzimidazol-2-ylacetonitril 99%
(2-Benzimidazolyl)acetonitrile

Purity: 99% CAS No.: 4414-88-4 Molecular Formula: C9H7N3 Formula Weight: 157.18 Odour: Odourless Melting Point: 206-211° Beilstein Reference: 128461 MDL No.: MFCD00005601

Formel: C₉H₇N₃
Schmelzpunkt: 206…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005601
CAS-Nummer: 4414-88-4
EINECS: 224-574-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR