Cell culture media, DMEM-Ham's F12
Lieferant: Biowest
Studies to determine the nutritional requirements of many cells have been in progress since Eagle's first reports. The major essential nutrients were identified, and work became focussed on the media requirements of individual cell types. Many media designed for these purposes are now available. Among the first of these media, developed initially to study hormonal requirements of cells in culture, was a mixture of DMEM and Ham's F12 medium, known as DMEM-Ham's F12.