Tetrakis(triphenylphosphin)palladium(0) ≥99%
Lieferant: Thermo Fisher Scientific
L00953.03
L00953.06
L00953.03EA
68.9
EUR
L00953.03
L00953.06
Tetrakis(triphenylphosphin)palladium(0) ≥99%
Tetrakis(triphenylphosphin)palladium(0)
Formel:
C₇₂H₆₀P₄Pd Schmelzpunkt: 103…107 °C Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00010012 CAS-Nummer: 14221-01-3 EINECS: 238-086-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...