Glutarimid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Piperidine-2,6-dione

L00968.06 L00968.14
L00968.06EA 42.1 EUR
L00968.06 L00968.14
Glutarimid ≥98%
Glutarimid

Purity: 98% CAS No.: 1121-89-7 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Odour: Nearly Odourless Melting Point: 153-157° Beilstein Reference: 110052 MDL No.: MFCD00006670

Formel: C₅H₇NO₂
Molecular Weight: 113.12 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006670
CAS-Nummer: 1121-89-7
EINECS: 214-340-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR