4-Nitro-2-(trifluormethyl)anilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Nitro-α,α,α-trifluor-o-toluidin, 2-Amino-5-nitro-α,α,α-trifluortoluol

L00977.06 L00977.14
L00977.06EA 24.2 EUR
L00977.06 L00977.14
4-Nitro-2-(trifluormethyl)anilin ≥98%
4-Nitro-2-(trifluormethyl)anilin

Purity: 98% CAS No.: 121-01-7 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 90-93° Beilstein Reference: 2121347 UN No.: 2811 MDL No.: MFCD00007365

Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206.12 g/mol
Schmelzpunkt: 90…93 °C
Dichte: 1.754 g/cm³ (–123.15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007365
CAS-Nummer: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR