4-Nitro-2-(trifluormethyl)anilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L00977.06
L00977.14
L00977.06EA
24.8
EUR
L00977.06
L00977.14
4-Nitro-2-(trifluormethyl)anilin ≥98%
4-Nitro-2-(trifluormethyl)anilin
Purity: 98%
CAS No.: 121-01-7
Molecular Formula: C7H5F3N2O2
Formula Weight: 206.13
Melting Point: 90-93°
Beilstein Reference: 2121347
UN No.: 2811
MDL No.: MFCD00007365
Formel:
C₇H₅F₃N₂O₂ Schmelzpunkt: 90…93 °C Dichte: 1,754 g/cm³ (–123.15 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007365 CAS-Nummer: 121-01-7 EINECS: 204-443-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...