2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.

Lieferant: Thermo Fisher Scientific

L01081.06 L01081.14
L01081.06EA 52.1 EUR
L01081.06 L01081.14
2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.
2-(8-Bromooctyloxy)tetrahydropyran

Purity: 90% CAS No.: 50816-20-1 Molecular Formula: C13H25BrO2 Formula Weight: 293.25 Flash Point: 110°(230°F) Refractive Index: 1.4770 Boiling Point: 130-132°/0.8mm Beilstein Reference: 1306644 MDL No.: MFCD00014648

Formel: C₁₃H₂₅BrO₂
Siedepunkt: 130…132 °C (0,8 mmHg)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014648
CAS-Nummer: 50816-20-1
EINECS: 256-786-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR