(Brommethyl)cyclopropan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L01122.03
L01122.06
L01122.14
AV13122.0100
L01122.03EA
46.5
EUR
L01122.03
L01122.06
L01122.14
ABCRAV13122.0100
(Brommethyl)cyclopropan ≥97%
(Brommethyl)cyclopropan
Purity: 97%
CAS No.: 7051-34-5
Molecular Formula: C4H7Br
Formula Weight: 135.00
Storage Temperature: 2°C to 8°C
Flash Point: 41°(105°F)
Refractive Index: 1.4770
Boiling Point: 111-112°
Beilstein Reference: 605296
UN No.: 1993
MDL No.: MFCD00001306
Formel:
C₄H₇Br Siedepunkt: 111…112 °C Dichte: 1,410 Flammpunkt: 41 °C (105 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001306 CAS-Nummer: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...