(Brommethyl)cyclopropan ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L01122.03 L01122.06 L01122.14 AV13122.0100
L01122.03EA 46.5 EUR
L01122.03 L01122.06 L01122.14 ABCRAV13122.0100
(Brommethyl)cyclopropan ≥97%
(Brommethyl)cyclopropan

Purity: 97% CAS No.: 7051-34-5 Molecular Formula: C4H7Br Formula Weight: 135.00 Storage Temperature: 2°C to 8°C Flash Point: 41°(105°F) Refractive Index: 1.4770 Boiling Point: 111-112° Beilstein Reference: 605296 UN No.: 1993 MDL No.: MFCD00001306

Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 111…112 °C
Dichte: 1.410
Flammpunkt: 41 °C (105 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR