Isochinolin-5-amin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Isoquinolin-5-amine, 5-Aminoisoquinoline
L01223.03
L01223.06
L01223.14
L01223.03EA
28.2
EUR
L01223.03
L01223.06
L01223.14
Isochinolin-5-amin 99%
Isochinolin-5-amin
Purity: 99%
CAS No.: 1125-60-6
Molecular Formula: C9H8N2
Formula Weight: 144.18
Melting Point: 126-131°
Beilstein Reference: 114465
Formel:
C₉H₈N₂ Schmelzpunkt: 126…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006907 CAS-Nummer: 1125-60-6 EINECS: 214-408-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...