3-Chlor-2-benzo[b]thiophencarbonsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L01257.03 L01257.06
L01257.03EA 39.4 EUR
L01257.03 L01257.06
3-Chlor-2-benzo[b]thiophencarbonsäure ≥97%
3-Chlor-2-benzo[b]thiophencarbonsäure

Purity: 97% CAS No.: 21211-22-3 Molecular Formula: C9H5ClO2S Formula Weight: 212.65 Melting Point: 265-267° Beilstein Reference: 1425602

Formel: C₉H₅ClO₂S
Schmelzpunkt: 265…267 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014579
CAS-Nummer: 21211-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR