1,3-Bis(trimethylsilyl)harnstoff ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N,N'-Bis(trimethylsilyl)urea

L01301.18 L01301.30
L01301.18EA 26.6 EUR
L01301.18 L01301.30
1,3-Bis(trimethylsilyl)harnstoff ≥98%
1,3-Bis(trimethylsilyl)harnstoff

Purity: 98+% CAS No.: 18297-63-7 Molecular Formula: C7H20N2OSi2 Formula Weight: 204.42 Melting Point: ca 230° dec. Beilstein Reference: 1937452

Formel: C₇H₂₀N₂OSi₂
Molecular Weight: 204.42 g/mol
Schmelzpunkt: ∼ 230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008260
CAS-Nummer: 18297-63-7
EINECS: 242-177-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR