3-(3-Methoxyphenyl)propionsäure ≥98%
Lieferant: Thermo Fisher Scientific
L01304.06
L01304.14
L01304.06EA
44.1
EUR
L01304.06
L01304.14
3-(3-Methoxyphenyl)propionsäure ≥98%
3-(3-Methoxyphenyl)propionsäure
Purity: 98+%
CAS No.: 10516-71-9
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 43-47°
Beilstein Reference: 1874422
Formel:
C₁₀H₁₂O₃ Schmelzpunkt: 43…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014027 CAS-Nummer: 10516-71-9 EINECS: 234-049-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...