3-(3-Methoxyphenyl)propionsäure ≥98%

Lieferant: Thermo Fisher Scientific

L01304.06 L01304.14
L01304.06EA 44.1 EUR
L01304.06 L01304.14
3-(3-Methoxyphenyl)propionsäure ≥98%
3-(3-Methoxyphenyl)propionsäure

Purity: 98+% CAS No.: 10516-71-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 43-47° Beilstein Reference: 1874422

Formel: C₁₀H₁₂O₃
Schmelzpunkt: 43…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014027
CAS-Nummer: 10516-71-9
EINECS: 234-049-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR